The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018117.11 | NP_060587.8 | 10 - 11 | 1716 | 85 |
XM_005269962.2 | XP_005270019.1 | 10 - 11 | 1433 | 85 | |
XM_005269963.2 | XP_005270020.1 | 10 - 11 | 1565 | 85 | |
XM_005269964.2 | XP_005270021.1 | 3 - 4 | 471 | 85 | |
XM_017016397.1 | XP_016871886.1 | 9 - 10 | 1404 | 85 | |
XM_017016398.1 | XP_016871887.1 | 9 - 10 | 1334 | 85 | |
XM_017016399.1 | XP_016871888.1 | 9 - 10 | 1278 | 85 | |
XM_017016400.1 | XP_016871889.1 | 10 - 11 | 1607 | 85 | |
GenBank mRNA | AB037772.2 | - | 10 - 11 | 1529 | 85 |
AF320223.1 | - | 10 - 11 | 1503 | 85 | |
AK001368.1 | - | 5 - 6 | 619 | 85 | |
AK299524.1 | - | 3 - 4 | 336 | 85 | |
AK315145.1 | - | 10 - 11 | 1494 | 85 | |
AL137699.1 | - | 8 - 9 | 1189 | 85 | |
BC040469.1 | - | 10 - 11 | 1533 | 85 | |
BC071564.1 | - | 10 - 11 | 1499 | 85 |