The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152754.2 | NP_689967.2 | 3 - 4 | 415 | 125 |
XM_011515960.1 | XP_011514262.1 | 5 - 6 | 725 | 125 | |
XM_011515961.2 | XP_011514263.1 | 3 - 4 | 250 | 125 | |
XM_017011873.1 | XP_016867362.1 | 4 - 5 | 914 | 125 | |
GenBank mRNA | AK309116.1 | - | 5 - 6 | 725 | 125 |
AY358937.1 | - | 4 - 5 | 481 | 125 |