Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCF7L2
Entrez Gene ID
6934
Gene Name
transcription factor 7 like 2
Gene Aliases
TCF-4, TCF4
Chromosome Location
Chr.10: 112950219 - 113167678 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146274.1 NP_001139746.1 11 - 12 1778 86
NM_001146283.1 NP_001139755.1 11 - 12 1850 86
NM_001146284.1 NP_001139756.1 10 - 11 1697 86
NM_001146285.1 NP_001139757.1 10 - 11 1709 86
NM_001146286.1 NP_001139758.1 10 - 11 1709 86
NM_001198525.1 NP_001185454.1 10 - 11 1724 86
NM_001198526.1 NP_001185455.1 10 - 11 1709 86
NM_001198527.1 NP_001185456.1 10 - 11 1697 86
NM_001198528.1 NP_001185457.1 10 - 11 1709 86
NM_001198529.1 NP_001185458.1 10 - 11 1709 86
NM_001198530.1 NP_001185459.1 9 - 10 1607 86
NM_001198531.1 NP_001185460.1 11 - 12 1778 86
NM_030756.4 NP_110383.2 10 - 11 1709 86
XM_005270071.1 XP_005270128.1 12 - 13 1968 86
XM_005270075.1 XP_005270132.1 12 - 13 1953 86
XM_005270077.1 XP_005270134.1 12 - 13 1968 86
XM_005270078.1 XP_005270135.1 12 - 13 1968 86
XM_005270079.1 XP_005270136.1 12 - 13 1953 86
XM_005270080.1 XP_005270137.1 11 - 12 1899 86
XM_005270084.1 XP_005270141.1 11 - 12 1809 86
XM_005270085.1 XP_005270142.1 11 - 12 1824 86
XM_005270086.1 XP_005270143.1 10 - 11 1755 86
XM_005270088.1 XP_005270145.1 10 - 11 1728 86
XM_005270089.1 XP_005270146.1 10 - 11 1755 86
XM_005270091.2 XP_005270148.1 12 - 13 1968 86
XM_005270092.1 XP_005270149.1 12 - 13 1968 86
XM_005270093.2 XP_005270150.1 12 - 13 1968 86
XM_005270094.2 XP_005270151.1 12 - 13 1965 86
XM_005270095.1 XP_005270152.1 12 - 13 1968 86
XM_005270096.2 XP_005270153.1 11 - 12 1809 86
XM_005270100.1 XP_005270157.1 11 - 12 1824 86
XM_005270101.2 XP_005270158.1 10 - 11 1755 86
XM_005270102.1 XP_005270159.1 11 - 12 1797 86
XM_005270103.1 XP_005270160.1 10 - 11 1755 86
XM_005270104.1 XP_005270161.1 10 - 11 1728 86
XM_011540109.1 XP_011538411.1 12 - 13 1968 86
XM_011540110.1 XP_011538412.1 11 - 12 1872 86
XM_011540111.1 XP_011538413.1 10 - 11 1740 86
XM_011540113.2 XP_011538415.1 12 - 13 1968 86
XM_011540116.1 XP_011538418.1 12 - 13 1968 86
XM_017016584.1 XP_016872073.1 12 - 13 1965 86
XM_017016585.1 XP_016872074.1 12 - 13 1956 86
XM_017016586.1 XP_016872075.1 12 - 13 1953 86
XM_017016587.1 XP_016872076.1 12 - 13 1941 86
XM_017016588.1 XP_016872077.1 11 - 12 1899 86
XM_017016589.1 XP_016872078.1 12 - 13 1968 86
XM_017016590.1 XP_016872079.1 11 - 12 1824 86
XM_017016591.1 XP_016872080.1 11 - 12 1812 86
XM_017016592.1 XP_016872081.1 11 - 12 1809 86
XM_017016593.1 XP_016872082.1 11 - 12 1809 86
XM_017016594.1 XP_016872083.1 10 - 11 1755 86
XM_017016595.1 XP_016872084.1 12 - 13 1953 86
XM_017016596.1 XP_016872085.1 11 - 12 1797 86
GenBank mRNA AB440195.1 - 11 - 12 1271 86
AB451266.1 - 10 - 11 1202 86
AK074705.1 - 2 - 3 224 86
AK225809.1 - 10 - 11 1713 86
AK296305.1 - 7 - 8 1074 86
AK299295.1 - 10 - 11 1239 86
AK301483.1 - 8 - 9 998 86
AK303684.1 - 7 - 8 791 86
AK315243.1 - 11 - 12 1516 86
AK316111.1 - 7 - 8 787 86
BC032656.1 - 10 - 11 1286 86
BX648364.1 - 11 - 12 1146 86
CR536574.1 - 10 - 11 1202 86
FJ010164.1 - 9 - 10 1097 86
FJ010165.1 - 10 - 11 1229 86
FJ010166.1 - 10 - 11 1310 86
FJ010167.1 - 10 - 11 1295 86
FJ010168.1 - 10 - 11 1287 86
FJ010169.1 - 10 - 11 1299 86
FJ010170.1 - 10 - 11 1295 86
FJ010172.1 - 10 - 11 1299 86
FJ010173.1 - 9 - 10 1097 86
FJ010174.1 - 10 - 11 1657 86
HM352838.1 - 10 - 11 1217 86
HM352839.1 - 10 - 11 1217 86
HM352840.1 - 11 - 12 1259 86
HM352841.1 - 11 - 12 1286 86
HM352842.1 - 10 - 11 1202 86
HM352843.1 - 10 - 11 1190 86
HM352844.1 - 11 - 12 1271 86
HM352845.1 - 11 - 12 1331 86
HM352846.1 - 11 - 12 1271 86
HM352847.1 - 10 - 11 1202 86
HM352848.1 - 10 - 11 1202 86
HM352849.1 - 10 - 11 1217 86
HM352850.1 - 11 - 12 1271 86
HM352851.1 - 11 - 12 1286 86
Y11306.2 - 10 - 11 1509 86

View Full Product Details