Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCF7L2
Entrez Gene ID
6934
Gene Name
transcription factor 7 like 2
Gene Aliases
TCF-4, TCF4
Chromosome Location
Chr.10: 112950219 - 113167678 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146274.1 NP_001139746.1 9 - 10 1512 143
NM_001146283.1 NP_001139755.1 9 - 10 1584 143
NM_001146284.1 NP_001139756.1 8 - 9 1431 143
NM_001146285.1 NP_001139757.1 8 - 9 1443 143
NM_001146286.1 NP_001139758.1 8 - 9 1443 143
NM_001198525.1 NP_001185454.1 8 - 9 1458 143
NM_001198526.1 NP_001185455.1 8 - 9 1443 143
NM_001198527.1 NP_001185456.1 8 - 9 1431 143
NM_001198528.1 NP_001185457.1 8 - 9 1443 143
NM_001198529.1 NP_001185458.1 8 - 9 1443 143
NM_001198530.1 NP_001185459.1 7 - 8 1341 143
NM_001198531.1 NP_001185460.1 9 - 10 1512 143
NM_030756.4 NP_110383.2 8 - 9 1443 143
XM_005270071.1 XP_005270128.1 10 - 11 1702 143
XM_005270075.1 XP_005270132.1 10 - 11 1687 143
XM_005270077.1 XP_005270134.1 10 - 11 1702 143
XM_005270078.1 XP_005270135.1 10 - 11 1702 143
XM_005270079.1 XP_005270136.1 10 - 11 1687 143
XM_005270080.1 XP_005270137.1 9 - 10 1633 143
XM_005270084.1 XP_005270141.1 9 - 10 1543 143
XM_005270085.1 XP_005270142.1 9 - 10 1558 143
XM_005270086.1 XP_005270143.1 8 - 9 1489 143
XM_005270088.1 XP_005270145.1 8 - 9 1462 143
XM_005270089.1 XP_005270146.1 8 - 9 1489 143
XM_005270091.2 XP_005270148.1 10 - 11 1702 143
XM_005270092.1 XP_005270149.1 10 - 11 1702 143
XM_005270093.2 XP_005270150.1 10 - 11 1702 143
XM_005270094.2 XP_005270151.1 10 - 11 1699 143
XM_005270095.1 XP_005270152.1 10 - 11 1702 143
XM_005270096.2 XP_005270153.1 9 - 10 1543 143
XM_005270100.1 XP_005270157.1 9 - 10 1558 143
XM_005270101.2 XP_005270158.1 8 - 9 1489 143
XM_005270102.1 XP_005270159.1 9 - 10 1531 143
XM_005270103.1 XP_005270160.1 8 - 9 1489 143
XM_005270104.1 XP_005270161.1 8 - 9 1462 143
XM_011540109.1 XP_011538411.1 10 - 11 1702 143
XM_011540110.1 XP_011538412.1 9 - 10 1606 143
XM_011540111.1 XP_011538413.1 8 - 9 1474 143
XM_011540113.2 XP_011538415.1 10 - 11 1702 143
XM_011540116.1 XP_011538418.1 10 - 11 1702 143
XM_017016584.1 XP_016872073.1 10 - 11 1699 143
XM_017016585.1 XP_016872074.1 10 - 11 1690 143
XM_017016586.1 XP_016872075.1 10 - 11 1687 143
XM_017016587.1 XP_016872076.1 10 - 11 1675 143
XM_017016588.1 XP_016872077.1 9 - 10 1633 143
XM_017016589.1 XP_016872078.1 10 - 11 1702 143
XM_017016590.1 XP_016872079.1 9 - 10 1558 143
XM_017016591.1 XP_016872080.1 9 - 10 1546 143
XM_017016592.1 XP_016872081.1 9 - 10 1543 143
XM_017016593.1 XP_016872082.1 9 - 10 1543 143
XM_017016594.1 XP_016872083.1 8 - 9 1489 143
XM_017016595.1 XP_016872084.1 10 - 11 1687 143
XM_017016596.1 XP_016872085.1 9 - 10 1531 143
GenBank mRNA AB440195.1 - 9 - 10 1005 143
AB451266.1 - 8 - 9 936 143
AK225809.1 - 8 - 9 1447 143
AK296305.1 - 5 - 6 808 143
AK299295.1 - 8 - 9 973 143
AK301483.1 - 6 - 7 732 143
AK303684.1 - 5 - 6 525 143
AK315243.1 - 9 - 10 1250 143
AK316111.1 - 5 - 6 521 143
BC032656.1 - 8 - 9 1020 143
BX648364.1 - 9 - 10 880 143
CR536574.1 - 8 - 9 936 143
FJ010164.1 - 7 - 8 831 143
FJ010165.1 - 8 - 9 963 143
FJ010166.1 - 8 - 9 1044 143
FJ010167.1 - 8 - 9 1029 143
FJ010168.1 - 8 - 9 1021 143
FJ010169.1 - 8 - 9 1033 143
FJ010170.1 - 8 - 9 1029 143
FJ010171.1 - 7 - 8 871 143
FJ010172.1 - 8 - 9 1033 143
FJ010173.1 - 7 - 8 831 143
FJ010174.1 - 8 - 9 1391 143
HM352838.1 - 8 - 9 951 143
HM352839.1 - 8 - 9 951 143
HM352840.1 - 9 - 10 993 143
HM352841.1 - 9 - 10 1020 143
HM352842.1 - 8 - 9 936 143
HM352843.1 - 8 - 9 924 143
HM352844.1 - 9 - 10 1005 143
HM352845.1 - 9 - 10 1065 143
HM352846.1 - 9 - 10 1005 143
HM352847.1 - 8 - 9 936 143
HM352848.1 - 8 - 9 936 143
HM352849.1 - 8 - 9 951 143
HM352850.1 - 9 - 10 1005 143
HM352851.1 - 9 - 10 1020 143
Y11306.2 - 8 - 9 1243 143

View Full Product Details