The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146285.1 | NP_001139757.1 | 11 - 12 | 1754 | 71 |
XM_005270077.1 | XP_005270134.1 | 13 - 14 | 2013 | 71 | |
XM_005270079.1 | XP_005270136.1 | 13 - 14 | 1998 | 71 | |
XM_011540116.1 | XP_011538418.1 | 13 - 14 | 2013 | 71 | |
XM_017016593.1 | XP_016872082.1 | 12 - 13 | 1854 | 71 | |
GenBank mRNA | AK225809.1 | - | 11 - 12 | 1758 | 71 |
FJ010174.1 | - | 11 - 12 | 1702 | 71 |