Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM184C
Entrez Gene ID
55751
Gene Name
transmembrane protein 184C
Gene Aliases
TMEM34
Chromosome Location
Chr.4: 147617371 - 147637273 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018241.2 NP_060711.2 6 - 7 1234 132
GenBank mRNA AK000169.1 - 3 - 4 387 132
AK001708.1 - 6 - 7 1177 132
BC046128.1 - 6 - 7 1236 132

View Full Product Details