The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018241.2 | NP_060711.2 | 7 - 8 | 1348 | 130 |
GenBank mRNA | AK000169.1 | - | 4 - 5 | 501 | 130 |
AK001708.1 | - | 7 - 8 | 1291 | 130 | |
BM712423.1 | - | 1 - 2 | 68 | 130 |