The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017639.3 | NP_060109.2 | 20 - 21 | 5423 | 70 |
XM_011532045.1 | XP_011530347.1 | 15 - 16 | 7211 | 70 | |
XM_011532046.2 | XP_011530348.1 | 14 - 15 | 4867 | 70 | |
XM_011532049.2 | XP_011530351.1 | 7 - 8 | 2717 | 70 | |
GenBank mRNA | AK095103.1 | - | 3 - 4 | 760 | 70 |
AY354497.1 | - | 20 - 21 | 5423 | 70 | |
BX647835.1 | - | 3 - 4 | 399 | 70 |