Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMO7
Entrez Gene ID
4008
Gene Name
LIM domain 7
Gene Aliases
FBX20, FBXO20, LOMP
Chromosome Location
Chr.13: 75620434 - 75859870 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306080.1 NP_001293009.1 22 - 23 3954 70
NM_005358.5 NP_005349.3 21 - 22 3998 70
NM_015842.2 NP_056667.2 19 - 20 3645 70
XM_011535075.2 XP_011533377.1 23 - 24 3842 70
XM_011535078.1 XP_011533380.1 22 - 23 3800 70
XM_011535080.2 XP_011533382.1 23 - 24 3761 70
XM_011535082.1 XP_011533384.1 22 - 23 3719 70
XM_011535086.1 XP_011533388.1 21 - 22 3515 70
XM_011535094.1 XP_011533396.1 16 - 17 2423 70
XM_011535095.1 XP_011533397.1 19 - 20 2594 70
XM_017020574.1 XP_016876063.1 23 - 24 3842 70
XM_017020575.1 XP_016876064.1 22 - 23 3800 70
XM_017020576.1 XP_016876065.1 22 - 23 3785 70
XM_017020577.1 XP_016876066.1 23 - 24 3761 70
XM_017020578.1 XP_016876067.1 21 - 22 3743 70
XM_017020579.1 XP_016876068.1 22 - 23 3719 70
XM_017020580.1 XP_016876069.1 22 - 23 3638 70
XM_017020581.1 XP_016876070.1 22 - 23 3764 70
XM_017020582.1 XP_016876071.1 22 - 23 3557 70
XM_017020583.1 XP_016876072.1 22 - 23 3557 70
XM_017020584.1 XP_016876073.1 21 - 22 3560 70
XM_017020585.1 XP_016876074.1 21 - 22 3356 70
XM_017020586.1 XP_016876075.1 21 - 22 3275 70
XM_017020587.1 XP_016876076.1 20 - 21 3233 70
XM_017020588.1 XP_016876077.1 20 - 21 3314 70
XM_017020589.1 XP_016876078.1 20 - 21 3437 70
XM_017020590.1 XP_016876079.1 20 - 21 3233 70
XM_017020591.1 XP_016876080.1 22 - 23 3925 70
XM_017020592.1 XP_016876081.1 22 - 23 3808 70
XM_017020593.1 XP_016876082.1 21 - 22 3721 70
XM_017020594.1 XP_016876083.1 21 - 22 3604 70
XM_017020595.1 XP_016876084.1 20 - 21 3598 70
XM_017020596.1 XP_016876085.1 20 - 21 2636 70
XM_017020597.1 XP_016876086.1 19 - 20 2606 70
XM_017020598.1 XP_016876087.1 20 - 21 3598 70
XM_017020599.1 XP_016876088.1 20 - 21 3439 70
XM_017020600.1 XP_016876089.1 20 - 21 2555 70
XM_017020601.1 XP_016876090.1 19 - 20 3316 70
XM_017020602.1 XP_016876091.1 19 - 20 3199 70
XM_017020603.1 XP_016876092.1 19 - 20 3316 70
XM_017020604.1 XP_016876093.1 19 - 20 2719 70
GenBank mRNA AB020665.2 - 19 - 20 3575 70
AF144237.1 - 10 - 11 1458 70
AF330045.1 - 21 - 22 3997 70
AK055465.1 - 14 - 15 2610 70
AK074265.1 - 6 - 7 447 70
AK092052.1 - 20 - 21 2917 70
AK304051.1 - 17 - 18 2536 70
FJ711162.1 - 22 - 23 3584 70

View Full Product Details