Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMO7
Entrez Gene ID
4008
Gene Name
LIM domain 7
Gene Aliases
FBX20, FBXO20, LOMP
Chromosome Location
Chr.13: 75620434 - 75859870 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306080.1 NP_001293009.1 6 - 7 836 122
NM_005358.5 NP_005349.3 7 - 8 1882 122
NM_015842.2 NP_056667.2 3 - 4 527 122
XM_011535075.2 XP_011533377.1 5 - 6 397 122
XM_011535078.1 XP_011533380.1 5 - 6 397 122
XM_011535080.2 XP_011533382.1 5 - 6 397 122
XM_011535082.1 XP_011533384.1 5 - 6 397 122
XM_011535086.1 XP_011533388.1 5 - 6 397 122
XM_011535094.1 XP_011533396.1 3 - 4 256 122
XM_011535095.1 XP_011533397.1 5 - 6 397 122
XM_017020574.1 XP_016876063.1 5 - 6 397 122
XM_017020575.1 XP_016876064.1 5 - 6 397 122
XM_017020576.1 XP_016876065.1 5 - 6 397 122
XM_017020577.1 XP_016876066.1 5 - 6 397 122
XM_017020578.1 XP_016876067.1 5 - 6 397 122
XM_017020579.1 XP_016876068.1 5 - 6 397 122
XM_017020580.1 XP_016876069.1 5 - 6 397 122
XM_017020581.1 XP_016876070.1 4 - 5 319 122
XM_017020582.1 XP_016876071.1 5 - 6 397 122
XM_017020583.1 XP_016876072.1 5 - 6 397 122
XM_017020584.1 XP_016876073.1 4 - 5 319 122
XM_017020585.1 XP_016876074.1 5 - 6 397 122
XM_017020586.1 XP_016876075.1 5 - 6 397 122
XM_017020587.1 XP_016876076.1 5 - 6 397 122
XM_017020588.1 XP_016876077.1 5 - 6 397 122
XM_017020589.1 XP_016876078.1 4 - 5 319 122
XM_017020590.1 XP_016876079.1 5 - 6 397 122
XM_017020591.1 XP_016876080.1 4 - 5 480 122
XM_017020592.1 XP_016876081.1 4 - 5 363 122
XM_017020593.1 XP_016876082.1 4 - 5 480 122
XM_017020594.1 XP_016876083.1 4 - 5 363 122
XM_017020595.1 XP_016876084.1 4 - 5 480 122
XM_017020596.1 XP_016876085.1 5 - 6 397 122
XM_017020597.1 XP_016876086.1 5 - 6 397 122
XM_017020598.1 XP_016876087.1 4 - 5 480 122
XM_017020599.1 XP_016876088.1 4 - 5 480 122
XM_017020600.1 XP_016876089.1 5 - 6 397 122
XM_017020601.1 XP_016876090.1 4 - 5 480 122
XM_017020602.1 XP_016876091.1 4 - 5 363 122
XM_017020603.1 XP_016876092.1 4 - 5 480 122
XM_017020604.1 XP_016876093.1 4 - 5 480 122
GenBank mRNA AB020665.2 - 3 - 4 457 122
AF330045.1 - 7 - 8 1881 122
AK092052.1 - 6 - 7 801 122
AK304051.1 - 4 - 5 369 122
FJ711162.1 - 6 - 7 466 122

View Full Product Details