Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMO7
Entrez Gene ID
4008
Gene Name
LIM domain 7
Gene Aliases
FBX20, FBXO20, LOMP
Chromosome Location
Chr.13: 75620434 - 75859870 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306080.1 NP_001293009.1 17 - 18 3598 65
NM_005358.5 NP_005349.3 16 - 17 3642 65
NM_015842.2 NP_056667.2 14 - 15 3289 65
XM_011535080.2 XP_011533382.1 17 - 18 3363 65
XM_011535082.1 XP_011533384.1 17 - 18 3363 65
XM_011535086.1 XP_011533388.1 16 - 17 3159 65
XM_017020577.1 XP_016876066.1 17 - 18 3363 65
XM_017020579.1 XP_016876068.1 17 - 18 3363 65
XM_017020582.1 XP_016876071.1 16 - 17 3159 65
XM_017020583.1 XP_016876072.1 16 - 17 3159 65
XM_017020586.1 XP_016876075.1 15 - 16 2877 65
XM_017020587.1 XP_016876076.1 15 - 16 2877 65
XM_017020589.1 XP_016876078.1 15 - 16 3081 65
XM_017020590.1 XP_016876079.1 15 - 16 2877 65
XM_017020595.1 XP_016876084.1 15 - 16 3242 65
XM_017020598.1 XP_016876087.1 15 - 16 3242 65
XM_017020600.1 XP_016876089.1 14 - 15 2157 65
XM_017020601.1 XP_016876090.1 14 - 15 2960 65
XM_017020602.1 XP_016876091.1 14 - 15 2843 65
XM_017020603.1 XP_016876092.1 14 - 15 2960 65
GenBank mRNA AB020665.2 - 14 - 15 3219 65
AF144237.1 - 5 - 6 1102 65
AF330045.1 - 16 - 17 3641 65
AK055465.1 - 9 - 10 2254 65
AK074265.1 - 1 - 2 91 65
AK092052.1 - 15 - 16 2561 65
FJ711162.1 - 17 - 18 3228 65

View Full Product Details