The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000069.2 | NP_000060.2 | 14 - 15 | 2289 | 73 |
XM_005245478.3 | XP_005245535.1 | 14 - 15 | 2176 | 73 | |
GenBank mRNA | BC133671.1 | - | 14 - 15 | 2133 | 73 |
L33798.1 | - | 14 - 15 | 2287 | 73 | |
U14413.1 | - | 1 - 2 | 67 | 73 |