The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000069.2 | NP_000060.2 | 27 - 28 | 3753 | 94 |
XM_005245478.3 | XP_005245535.1 | 27 - 28 | 3640 | 94 | |
GenBank mRNA | BC133671.1 | - | 27 - 28 | 3597 | 94 |
L33798.1 | - | 27 - 28 | 94 |