The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000069.2 | NP_000060.2 | 37 - 38 | 4777 | 77 |
XM_005245478.3 | XP_005245535.1 | 36 - 37 | 4607 | 77 | |
GenBank mRNA | BC133671.1 | - | 37 - 38 | 4621 | 77 |
L33798.1 | - | 37 - 38 | 4775 | 77 |