Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACNA1S
Entrez Gene ID
779
Gene Name
calcium voltage-gated channel subunit alpha1 S
Gene Aliases
CACNL1A3, CCHL1A3, Cav1.1, HOKPP, HOKPP1, MHS5, TTPP1, hypoPP
Chromosome Location
Chr.1: 201039509 - 201112566 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000069.2 NP_000060.2 41 - 42 5367 91
XM_005245478.3 XP_005245535.1 40 - 41 5197 91
GenBank mRNA BC133671.1 - 41 - 42 5211 91
L33798.1 - 41 - 42 91

View Full Product Details