Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SGIP1
Entrez Gene ID
84251
Gene Name
SH3 domain GRB2 like endophilin interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.1: 66533272 - 66751139 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308203.1 NP_001295132.1 7 - 8 610 67
XM_005271270.4 XP_005271327.1 7 - 8 949 67
XM_006710973.2 XP_006711036.1 7 - 8 702 67
XM_006710974.2 XP_006711037.1 6 - 7 618 67
XM_011542293.1 XP_011540595.1 6 - 7 618 67
XM_017002509.1 XP_016857998.1 7 - 8 690 67
XM_017002510.1 XP_016857999.1 8 - 9 813 67
XM_017002515.1 XP_016858004.1 6 - 7 618 67
XM_017002516.1 XP_016858005.1 7 - 8 741 67
XM_017002524.1 XP_016858013.1 8 - 9 806 67
XM_017002525.1 XP_016858014.1 8 - 9 805 67
XM_017002526.1 XP_016858015.1 7 - 8 690 67
XM_017002527.1 XP_016858016.1 8 - 9 714 67
XM_017002528.1 XP_016858017.1 7 - 8 734 67
XM_017002531.1 XP_016858020.1 7 - 8 702 67
XM_017002533.1 XP_016858022.1 8 - 9 804 67
XM_017002534.1 XP_016858023.1 6 - 7 618 67
XM_017002535.1 XP_016858024.1 7 - 8 732 67
XM_017002537.1 XP_016858026.1 7 - 8 731 67
GenBank mRNA AK090697.1 - 1 - 2 167 67
BX640813.1 - 7 - 8 587 67
CR749541.1 - 7 - 8 804 67

View Full Product Details