The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015547.3 | NP_056362.1 | 2 - 3 | 415 | 86 |
NM_147161.3 | NP_671517.1 | 2 - 3 | 415 | 86 | |
GenBank mRNA | AB014607.1 | - | 2 - 3 | 310 | 86 |
AF416921.1 | - | 2 - 3 | 257 | 86 | |
AF416922.1 | - | 2 - 3 | 261 | 86 | |
AK023937.1 | - | 2 - 3 | 321 | 86 | |
AK223570.1 | - | 2 - 3 | 321 | 86 | |
BC001517.2 | - | 3 - 4 | 482 | 86 | |
BC093844.1 | - | 2 - 3 | 281 | 86 | |
BC093846.1 | - | 2 - 3 | 281 | 86 | |
DA121227.1 | - | 2 - 3 | 415 | 86 |