Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERINC2
Entrez Gene ID
347735
Gene Name
serine incorporator 2
Gene Aliases
FKSG84, PRO0899, TDE2, TDE2L
Chromosome Location
Chr.1: 31409565 - 31434680 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199039.1 NP_001185968.1 2 - 3 999 105
GenBank mRNA AK093784.1 - 2 - 3 1007 105

View Full Product Details