Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACAT1
Entrez Gene ID
38
Gene Name
acetyl-CoA acetyltransferase 1
Gene Aliases
ACAT, MAT, T2, THIL
Chromosome Location
Chr.11: 108121531 - 108148168 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000019.3 NP_000010.1 1 - 2 151 163
GenBank mRNA D90228.1 - 1 - 2 151 163

View Full Product Details