The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153705.4 | NP_714916.3 | 1 - 2 | 269 | 90 |
XM_011542623.2 | XP_011540925.1 | 1 - 2 | 236 | 90 | |
GenBank mRNA | AF533708.1 | - | 1 - 2 | 228 | 90 |
AK313534.1 | - | 1 - 2 | 131 | 90 | |
BC036526.2 | - | 1 - 2 | 223 | 90 | |
DN999478.1 | - | 1 - 2 | 277 | 90 |