Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KDELC2
Entrez Gene ID
143888
Gene Name
KDEL motif containing 2
Gene Aliases
-
Chromosome Location
Chr.11: 108472106 - 108498432 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153705.4 NP_714916.3 3 - 4 753 124
XM_011542621.2 XP_011540923.1 3 - 4 726 124
GenBank mRNA AF533708.1 - 3 - 4 712 124
AK125519.1 - 2 - 3 535 124
AK313534.1 - 3 - 4 615 124
AY358616.1 - 2 - 3 822 124
BC036526.2 - 3 - 4 707 124

View Full Product Details