The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025004.2 | NP_079280.2 | 7 - 8 | 1153 | 106 |
XM_017018346.1 | XP_016873835.1 | 7 - 8 | 1153 | 106 | |
XM_017018347.1 | XP_016873836.1 | 7 - 8 | 1153 | 106 | |
XM_017018348.1 | XP_016873837.1 | 7 - 8 | 1153 | 106 | |
XM_017018349.1 | XP_016873838.1 | 6 - 7 | 2294 | 106 | |
XM_017018350.1 | XP_016873839.1 | 7 - 8 | 1153 | 106 | |
XM_017018351.1 | XP_016873840.1 | 7 - 8 | 1153 | 106 | |
XM_017018352.1 | XP_016873841.1 | 6 - 7 | 803 | 106 | |
XM_017018353.1 | XP_016873842.1 | 7 - 8 | 1153 | 106 | |
GenBank mRNA | AK023277.1 | - | 7 - 8 | 1153 | 106 |
BC018540.1 | - | 7 - 8 | 1067 | 106 |