The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142685.1 | NP_001136157.1 | 17 - 18 | 1940 | 102 |
NM_014715.3 | NP_055530.2 | 8 - 9 | 1272 | 102 | |
XM_005271736.4 | XP_005271793.2 | 18 - 19 | 2970 | 102 | |
XM_011543072.2 | XP_011541374.1 | 18 - 19 | 2172 | 102 | |
XM_011543073.2 | XP_011541375.2 | 18 - 19 | 1908 | 102 | |
XM_011543075.2 | XP_011541377.1 | 9 - 10 | 1118 | 102 | |
XM_017018595.1 | XP_016874084.1 | 18 - 19 | 1889 | 102 | |
XM_017018596.1 | XP_016874085.1 | 16 - 17 | 2166 | 102 | |
GenBank mRNA | AB018255.2 | - | 8 - 9 | 1193 | 102 |
AB088416.1 | - | 8 - 9 | 1193 | 102 | |
AL833062.2 | - | 16 - 17 | 1742 | 102 | |
AY194287.1 | - | 8 - 9 | 1196 | 102 | |
BC104898.1 | - | 7 - 8 | 927 | 102 | |
BC113429.1 | - | 7 - 8 | 927 | 102 | |
EF127492.1 | - | 17 - 18 | 1940 | 102 |