The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142685.1 | NP_001136157.1 | 8 - 9 | 845 | 83 |
XM_005271736.4 | XP_005271793.2 | 9 - 10 | 1875 | 83 | |
XM_011543072.2 | XP_011541374.1 | 9 - 10 | 1077 | 83 | |
XM_011543073.2 | XP_011541375.2 | 9 - 10 | 813 | 83 | |
XM_011543075.2 | XP_011541377.1 | 1 - 2 | 105 | 83 | |
XM_017018595.1 | XP_016874084.1 | 9 - 10 | 794 | 83 | |
XM_017018596.1 | XP_016874085.1 | 7 - 8 | 1071 | 83 | |
XM_017018597.1 | XP_016874086.1 | 9 - 10 | 1884 | 83 | |
GenBank mRNA | AL833062.2 | - | 7 - 8 | 647 | 83 |
BC000277.1 | - | 1 - 2 | 91 | 83 | |
BC051236.1 | - | 3 - 4 | 301 | 83 | |
BC065371.1 | - | 3 - 4 | 301 | 83 | |
EF127492.1 | - | 8 - 9 | 845 | 83 |