The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142685.1 | NP_001136157.1 | 15 - 16 | 1587 | 135 |
NM_014715.3 | NP_055530.2 | 6 - 7 | 919 | 135 | |
XM_005271736.4 | XP_005271793.2 | 16 - 17 | 2617 | 135 | |
XM_011543072.2 | XP_011541374.1 | 16 - 17 | 1819 | 135 | |
XM_011543073.2 | XP_011541375.2 | 16 - 17 | 1555 | 135 | |
XM_011543075.2 | XP_011541377.1 | 7 - 8 | 765 | 135 | |
XM_017018595.1 | XP_016874084.1 | 16 - 17 | 1536 | 135 | |
XM_017018596.1 | XP_016874085.1 | 14 - 15 | 1813 | 135 | |
XM_017018597.1 | XP_016874086.1 | 16 - 17 | 2626 | 135 | |
GenBank mRNA | AB018255.2 | - | 6 - 7 | 840 | 135 |
AB079856.1 | - | 6 - 7 | 919 | 135 | |
AB088416.1 | - | 6 - 7 | 840 | 135 | |
AL833062.2 | - | 14 - 15 | 1389 | 135 | |
AY194287.1 | - | 6 - 7 | 843 | 135 | |
BC000277.1 | - | 8 - 9 | 833 | 135 | |
BC104898.1 | - | 5 - 6 | 574 | 135 | |
BC113429.1 | - | 5 - 6 | 574 | 135 | |
EF127492.1 | - | 15 - 16 | 1587 | 135 |