The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139025.4 | NP_620594.1 | 26 - 27 | 4158 | 61 |
NM_139026.4 | NP_620595.1 | 26 - 27 | 3897 | 61 | |
NM_139027.4 | NP_620596.2 | 26 - 27 | 3990 | 61 | |
NR_024514.2 | 16 - 17 | 2381 | 61 | ||
XM_011518176.2 | XP_011516478.1 | 19 - 20 | 2810 | 61 | |
XM_011518178.1 | XP_011516480.1 | 16 - 17 | 2484 | 61 | |
XM_011518179.1 | XP_011516481.1 | 16 - 17 | 2527 | 61 | |
XM_017014232.1 | XP_016869721.1 | 26 - 27 | 4369 | 61 | |
XM_017014233.1 | XP_016869722.1 | 26 - 27 | 3770 | 61 | |
XM_017014234.1 | XP_016869723.1 | 20 - 21 | 2958 | 61 | |
GenBank mRNA | AB069698.2 | - | 26 - 27 | 4158 | 61 |
AF414401.1 | - | 26 - 27 | 3775 | 61 | |
AJ305314.1 | - | 26 - 27 | 3573 | 61 | |
AJ420810.1 | - | 26 - 27 | 3666 | 61 | |
AK124983.1 | - | 4 - 5 | 735 | 61 | |
AL136809.1 | - | 2 - 3 | 298 | 61 | |
AY055376.1 | - | 26 - 27 | 3822 | 61 | |
AY358118.1 | - | 19 - 20 | 2926 | 61 |