The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139025.4 | NP_620594.1 | 9 - 10 | 1538 | 96 |
NM_139026.4 | NP_620595.1 | 9 - 10 | 1445 | 96 | |
NM_139027.4 | NP_620596.2 | 9 - 10 | 1538 | 96 | |
XM_011518176.2 | XP_011516478.1 | 2 - 3 | 190 | 96 | |
XM_011518179.1 | XP_011516481.1 | 1 - 2 | 92 | 96 | |
XM_017014232.1 | XP_016869721.1 | 9 - 10 | 1749 | 96 | |
XM_017014233.1 | XP_016869722.1 | 9 - 10 | 1150 | 96 | |
XM_017014234.1 | XP_016869723.1 | 3 - 4 | 338 | 96 | |
XM_017014235.1 | XP_016869724.1 | 9 - 10 | 2319 | 96 | |
GenBank mRNA | AB069698.2 | - | 9 - 10 | 1538 | 96 |
AF414401.1 | - | 9 - 10 | 1155 | 96 | |
AJ011374.1 | - | 3 - 4 | 728 | 96 | |
AJ305314.1 | - | 9 - 10 | 1121 | 96 | |
AJ420810.1 | - | 9 - 10 | 1214 | 96 | |
AK292439.1 | - | 8 - 9 | 1186 | 96 | |
AY055376.1 | - | 9 - 10 | 1202 | 96 | |
AY358118.1 | - | 2 - 3 | 306 | 96 | |
BC103828.1 | - | 7 - 8 | 912 | 96 | |
BC103829.1 | - | 7 - 8 | 912 | 96 |