Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ADAMTS13
- Entrez Gene ID
-
11093
- Gene Name
- ADAM metallopeptidase with thrombospondin type 1 motif 13
- Gene Aliases
-
ADAM-TS13, ADAMTS-13, C9orf8, VWFCP, vWF-CP
- Chromosome Location
-
Chr.9: 133414339 - 133459403 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_024514.2 |
|
1 - 2 |
298 |
70 |
GenBank mRNA |
DB069518.1 |
- |
1 - 2 |
301 |
70 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB091741.1 |
- |
1 - 2 |
282 |
|
Chr. 9 - 133414355 - 133425976 [+] on Build GRCh38 |
70 |
GenBankmRNA |
BI831048.1 |
- |
1 - 2 |
278 |
|
Chr. 9 - 133414358 - 133426261 [+] on Build GRCh38 |
70 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
serine/threonine kinase like domain containing 1 |
STKLD1 |
C9orf96, SgK071, Sk521 |
-8.2kb
|
Sense |
Chr. - 133376413 - 133406096 on Build GRCh38 |
Downstream |
calcium channel flower domain containing 1 |
CACFD1 |
C9orf7, D9S2135, FLOWER |
36.8kb
|
Sense |
Chr. - 133459965 - 133470787 on Build GRCh38 |
View Full Product Details