Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCSK7
Entrez Gene ID
9159
Gene Name
proprotein convertase subtilisin/kexin type 7
Gene Aliases
LPC, PC7, PC8, SPC7
Chromosome Location
Chr.11: 117205071 - 117232525 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004716.3 NP_004707.2 1 - 2 499 112
GenBank mRNA AK312429.1 - 1 - 2 74 112
BC010696.1 - 1 - 2 69 112
DA126121.1 - 1 - 2 499 112

View Full Product Details