Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCSK7
Entrez Gene ID
9159
Gene Name
proprotein convertase subtilisin/kexin type 7
Gene Aliases
LPC, PC7, PC8, SPC7
Chromosome Location
Chr.11: 117205071 - 117232525 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004716.3 NP_004707.2 2 - 3 615 62
GenBank mRNA AK222907.1 - 2 - 3 155 62
AK312429.1 - 2 - 3 190 62
BC006357.2 - 2 - 3 133 62
BC010696.1 - 2 - 3 185 62
U33849.1 - 2 - 3 135 62

View Full Product Details