The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004716.3 | NP_004707.2 | 11 - 12 | 2059 | 85 |
XM_006718940.3 | XP_006719003.1 | 4 - 5 | 818 | 85 | |
XM_017018546.1 | XP_016874035.1 | 6 - 7 | 587 | 85 | |
GenBank mRNA | AK097207.1 | - | 3 - 4 | 1234 | 85 |
AK222907.1 | - | 11 - 12 | 1599 | 85 | |
AK312429.1 | - | 11 - 12 | 1634 | 85 | |
BC006357.2 | - | 11 - 12 | 1577 | 85 | |
BC010696.1 | - | 11 - 12 | 1629 | 85 | |
BT006870.1 | - | 9 - 10 | 1428 | 85 | |
U33849.1 | - | 11 - 12 | 1579 | 85 | |
U40623.1 | - | 9 - 10 | 1449 | 85 |