The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166691.1 | NP_001160163.1 | 17 - 18 | 2166 | 84 |
NM_003318.4 | NP_003309.2 | 17 - 18 | 2157 | 84 | |
XM_011536099.2 | XP_011534401.1 | 17 - 18 | 2131 | 84 | |
XM_011536100.2 | XP_011534402.1 | 17 - 18 | 2154 | 84 | |
XM_017011242.1 | XP_016866731.1 | 18 - 19 | 2686 | 84 | |
GenBank mRNA | AK292460.1 | - | 17 - 18 | 2129 | 84 |
AK315696.1 | - | 17 - 18 | 2074 | 84 | |
AX775733.1 | - | 17 - 18 | 2129 | 84 | |
BC000633.2 | - | 17 - 18 | 2120 | 84 | |
BC032858.2 | - | 17 - 18 | 2122 | 84 | |
X70500.1 | - | 4 - 5 | 579 | 84 |