The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166691.1 | NP_001160163.1 | 19 - 20 | 2418 | 155 |
NM_003318.4 | NP_003309.2 | 19 - 20 | 2409 | 155 | |
XM_011536099.2 | XP_011534401.1 | 19 - 20 | 2383 | 155 | |
XM_011536100.2 | XP_011534402.1 | 19 - 20 | 2406 | 155 | |
XM_017011242.1 | XP_016866731.1 | 20 - 21 | 2938 | 155 | |
GenBank mRNA | AK292460.1 | - | 19 - 20 | 2381 | 155 |
AK315696.1 | - | 19 - 20 | 2326 | 155 | |
AX775733.1 | - | 19 - 20 | 2381 | 155 | |
BC000633.2 | - | 19 - 20 | 2372 | 155 | |
BC032858.2 | - | 19 - 20 | 2374 | 155 |