The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143850.2 | NP_001137322.1 | 11 - 12 | 1436 | 164 |
NM_024809.4 | NP_079085.2 | 11 - 12 | 1439 | 164 | |
XM_006719605.3 | XP_006719668.1 | 11 - 12 | 1439 | 164 | |
XM_017019975.1 | XP_016875464.1 | 10 - 11 | 1314 | 164 | |
GenBank mRNA | AK056924.1 | - | 11 - 12 | 1437 | 164 |
AK292153.1 | - | 11 - 12 | 1436 | 164 | |
BC009112.1 | - | 11 - 12 | 1408 | 164 | |
DQ278870.1 | - | 11 - 12 | 1437 | 164 |