Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
BDH2
Entrez Gene ID
56898
Gene Name
3-hydroxybutyrate dehydrogenase, type 2
Gene Aliases
DHRS6, EFA6R, PRO20933, SDR15C1, UCPA-OR, UNQ6308
Chromosome Location
Chr.4: 103077618 - 103099867 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020139.3 NP_064524.3 7 - 7 589 182
XM_006714274.3 XP_006714337.1 8 - 8 626 182
XM_011532127.2 XP_011530429.1 8 - 8 668 182
XM_011532128.2 XP_011530430.1 7 - 7 626 182
XM_017008461.1 XP_016863950.1 8 - 8 665 182
XM_017008462.1 XP_016863951.1 7 - 7 623 182
GenBank mRNA AF164790.1 - 7 - 7 553 182
AK023323.1 - 6 - 6 183
AK128463.1 - 7 - 7 2304 182
AK300626.1 - 7 - 7 434 182
BC001953.1 - 7 - 7 582 182
BC037277.2 - 6 - 6 1568 182
BC095414.1 - 7 - 7 557 182

View Full Product Details