The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006714274.3 | XP_006714337.1 | 7 - 8 | 564 | 75 |
XM_011532127.2 | XP_011530429.1 | 7 - 8 | 606 | 75 | |
XM_017008461.1 | XP_016863950.1 | 7 - 8 | 603 | 75 |