The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152744.3 | NP_689957.3 | 24 - 25 | 3721 | 74 |
XM_011515188.1 | XP_011513490.1 | 18 - 19 | 2559 | 74 | |
XM_011515189.1 | XP_011513491.1 | 18 - 19 | 2559 | 74 | |
XM_011515190.1 | XP_011513492.1 | 18 - 19 | 2559 | 74 | |
XM_017011837.1 | XP_016867326.1 | 18 - 19 | 2559 | 74 | |
XM_017011838.1 | XP_016867327.1 | 18 - 19 | 2559 | 74 | |
XM_017011839.1 | XP_016867328.1 | 18 - 19 | 2559 | 74 | |
XM_017011840.1 | XP_016867329.1 | 18 - 19 | 2559 | 74 | |
GenBank mRNA | AK074083.1 | - | 10 - 11 | 1417 | 74 |
AY310398.1 | - | 24 - 25 | 3721 | 74 |