The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079653.1 | NP_001073121.1 | 3 - 4 | 709 | 80 |
NM_152744.3 | NP_689957.3 | 29 - 30 | 4564 | 80 | |
XM_011515188.1 | XP_011513490.1 | 23 - 24 | 3402 | 80 | |
XM_011515189.1 | XP_011513491.1 | 23 - 24 | 3402 | 80 | |
XM_011515190.1 | XP_011513492.1 | 23 - 24 | 3402 | 80 | |
XM_017011837.1 | XP_016867326.1 | 23 - 24 | 3402 | 80 | |
XM_017011838.1 | XP_016867327.1 | 23 - 24 | 3402 | 80 | |
XM_017011839.1 | XP_016867328.1 | 23 - 24 | 3402 | 80 | |
XM_017011840.1 | XP_016867329.1 | 23 - 24 | 3402 | 80 | |
GenBank mRNA | AK074083.1 | - | 15 - 16 | 2260 | 80 |
AL833354.1 | - | 3 - 4 | 706 | 80 | |
AY310398.1 | - | 29 - 30 | 4564 | 80 |