The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079653.1 | NP_001073121.1 | 14 - 15 | 2110 | 63 |
NM_152744.3 | NP_689957.3 | 40 - 41 | 5965 | 63 | |
XM_011515188.1 | XP_011513490.1 | 34 - 35 | 4806 | 63 | |
XM_011515189.1 | XP_011513491.1 | 34 - 35 | 4803 | 63 | |
XM_017011837.1 | XP_016867326.1 | 34 - 35 | 4806 | 63 | |
GenBank mRNA | AK074077.1 | - | 7 - 8 | 905 | 63 |
AK074083.1 | - | 25 - 26 | 3601 | 63 | |
AL833354.1 | - | 14 - 15 | 2107 | 63 | |
AY310398.1 | - | 40 - 41 | 5965 | 63 |