Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYRIP
Entrez Gene ID
25924
Gene Name
myosin VIIA and Rab interacting protein
Gene Aliases
SLAC2-C, SLAC2C
Chromosome Location
Chr.3: 39808914 - 40260321 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284423.1 NP_001271352.1 14 - 15 2465 66
NM_001284424.1 NP_001271353.1 13 - 14 2459 66
NM_001284425.1 NP_001271354.1 13 - 14 2517 66
NM_001284426.1 NP_001271355.1 12 - 13 2275 66
NM_015460.3 NP_056275.2 14 - 15 2654 66
NR_104316.1 14 - 15 2617 66
XM_011533575.1 XP_011531877.1 14 - 15 2626 66
GenBank mRNA AB083783.1 - 13 - 14 2367 66
AF396687.1 - 14 - 15 2503 66
AK125334.1 - 12 - 13 2141 66
AK126013.1 - 13 - 14 2413 66
AK294714.1 - 12 - 13 2265 66
AK312412.1 - 14 - 15 2464 66
AK316068.1 - 12 - 13 2275 66
AL050090.1 - 2 - 3 273 66
BC092512.1 - 14 - 15 2653 66
BC109311.1 - 13 - 14 2345 66
BC109312.1 - 14 - 15 2445 66

View Full Product Details