Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANO7
Entrez Gene ID
50636
Gene Name
anoctamin 7
Gene Aliases
D-TMPP, DTMPP, IPCA-5, IPCA5, NGEP, PCANAP5, PCANAP5L, TMEM16G
Chromosome Location
Chr.2: 241188509 - 241239602 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001891.3 NP_001001891.2 18 - 19 2088 76
XM_011511263.2 XP_011509565.1 18 - 19 2036 76
XM_011511267.2 XP_011509569.1 18 - 19 2122 76
XM_017004230.1 XP_016859719.1 17 - 18 2017 76
GenBank mRNA AY617079.1 - 18 - 19 2089 76

View Full Product Details