The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198608.1 | NP_001185537.1 | 5 - 6 | 837 | 69 |
NM_001198609.1 | NP_001185538.1 | 5 - 6 | 837 | 69 | |
NM_001198614.1 | NP_001185543.1 | 6 - 7 | 840 | 69 | |
NM_001198615.1 | NP_001185544.1 | 5 - 6 | 570 | 69 | |
NM_001198618.1 | NP_001185547.1 | 5 - 6 | 700 | 69 | |
NM_001198619.1 | NP_001185548.1 | 5 - 6 | 573 | 69 | |
NM_003980.4 | NP_003971.1 | 5 - 6 | 933 | 69 | |
XM_006715598.2 | XP_006715661.1 | 6 - 7 | 1358 | 69 | |
XM_006715599.2 | XP_006715662.1 | 6 - 7 | 1127 | 69 | |
XM_006715600.2 | XP_006715663.1 | 6 - 7 | 1127 | 69 | |
XM_006715601.3 | XP_006715664.1 | 5 - 6 | 1129 | 69 | |
XM_011536243.2 | XP_011534545.1 | 5 - 6 | 1728 | 69 | |
GenBank mRNA | AK295749.1 | - | 5 - 6 | 565 | 69 |
AK296483.1 | - | 5 - 6 | 700 | 69 | |
AK296556.1 | - | 5 - 6 | 699 | 69 | |
AK299355.1 | - | 5 - 6 | 570 | 69 | |
AK307873.1 | - | 5 - 6 | 676 | 69 | |
AK311114.1 | - | 3 - 4 | 199 | 69 | |
AK311234.1 | - | 6 - 7 | 717 | 69 | |
AK311236.1 | - | 6 - 7 | 675 | 69 | |
AK312789.1 | - | 5 - 6 | 533 | 69 | |
AK316062.1 | - | 5 - 6 | 573 | 69 | |
AK316529.1 | - | 6 - 7 | 706 | 69 | |
BC025777.1 | - | 5 - 6 | 529 | 69 | |
X73882.1 | - | 5 - 6 | 622 | 69 |