Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCAPH
Entrez Gene ID
23397
Gene Name
non-SMC condensin I complex subunit H
Gene Aliases
BRRN1, CAP-H
Chromosome Location
Chr.2: 96335741 - 96377095 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281710.1 NP_001268639.1 6 - 7 782 70
NM_001281711.1 NP_001268640.1 6 - 7 743 70
NM_001281712.1 NP_001268641.1 5 - 6 562 70
NM_015341.4 NP_056156.2 6 - 7 815 70
XM_005263908.3 XP_005263965.1 6 - 7 755 70
XM_006712388.3 XP_006712451.1 6 - 7 755 70
GenBank mRNA AK291053.1 - 6 - 7 756 70
AK299248.1 - 6 - 7 684 70
AK303725.1 - 5 - 6 507 70
AK308050.1 - 6 - 7 722 70
AK310649.1 - 6 - 7 683 70
BC024211.2 - 6 - 7 769 70
D38553.1 - 6 - 7 745 70

View Full Product Details