The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001971.5 | NP_001962.3 | 7 - 8 | 806 | 69 |
GenBank mRNA | AF120493.1 | - | 7 - 8 | 806 | 69 |
BC069454.1 | - | 7 - 8 | 806 | 69 | |
BC075091.2 | - | 7 - 8 | 787 | 69 |