The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_026676.1 | 1 - 1 | 860 | 66 | |
GenBank mRNA | BC026177.1 | - | 1 - 1 | 669 | 66 |