Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RARS
Entrez Gene ID
5917
Gene Name
arginyl-tRNA synthetase
Gene Aliases
ArgRS, DALRD1, HLD9
Chromosome Location
Chr.5: 168486458 - 168519306 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002887.3 NP_002878.2 5 - 6 627 108
GenBank mRNA AK222797.1 - 5 - 6 603 108
AK302161.1 - 4 - 5 485 108
AK314795.1 - 5 - 6 615 108
BC000528.2 - 5 - 6 605 108
BC014619.2 - 5 - 6 605 108
BT007394.1 - 5 - 6 586 108
S80343.1 - 5 - 6 613 108

View Full Product Details