The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033503.3 | NP_277038.1 | 1 - 2 | 81 | 70 |
GenBank mRNA | AK024472.1 | - | 1 - 2 | 91 | 70 |
BC069505.1 | - | 1 - 2 | 65 | 70 | |
CB108463.1 | - | 3 - 4 | 316 | 70 |