The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145290.1 | NP_001138762.1 | 12 - 13 | 1380 | 66 |
NM_198277.2 | NP_938018.1 | 12 - 13 | 1380 | 66 | |
GenBank mRNA | AK074207.1 | - | 12 - 13 | 1380 | 66 |
AK290314.1 | - | 12 - 13 | 1192 | 66 | |
BC051314.1 | - | 12 - 13 | 1161 | 66 |