The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270364.1 | NP_001257293.1 | 21 - 22 | 2698 | 74 |
NM_001270365.1 | NP_001257294.1 | 19 - 20 | 2497 | 74 | |
NM_012190.3 | NP_036322.2 | 21 - 22 | 2800 | 74 | |
NR_072979.1 | 19 - 20 | 2549 | 74 | ||
XM_006713481.2 | XP_006713544.1 | 21 - 22 | 2539 | 74 | |
XM_011512355.1 | XP_011510657.1 | 21 - 22 | 2532 | 74 | |
XM_017005613.1 | XP_016861102.1 | 19 - 20 | 2701 | 74 | |
XM_017005614.1 | XP_016861103.1 | 19 - 20 | 2317 | 74 | |
GenBank mRNA | AB209299.1 | - | 21 - 22 | 2607 | 74 |
AF052732.1 | - | 21 - 22 | 2560 | 74 | |
AK124908.1 | - | 4 - 5 | 1315 | 74 | |
AK222535.1 | - | 21 - 22 | 2582 | 74 | |
AK222694.1 | - | 21 - 22 | 2582 | 74 | |
AK294392.1 | - | 19 - 20 | 2281 | 74 | |
BC027241.1 | - | 19 - 20 | 2322 | 74 | |
CR749807.1 | - | 21 - 22 | 2698 | 74 |