The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000019.3 | NP_000010.1 | 8 - 9 | 907 | 80 |
XM_006718835.3 | XP_006718898.1 | 8 - 9 | 890 | 80 | |
XM_017017681.1 | XP_016873170.1 | 9 - 10 | 986 | 80 | |
XM_017017682.1 | XP_016873171.1 | 7 - 8 | 770 | 80 | |
XM_017017683.1 | XP_016873172.1 | 8 - 9 | 863 | 80 | |
GenBank mRNA | AK312574.1 | - | 8 - 9 | 874 | 80 |
D90228.1 | - | 8 - 9 | 907 | 80 |